Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM262P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: BM-2. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: HL60 cells. Bone marrow, lymph node or tonsil. Cellular Staining: Cytoplasmic, nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C.
Catalog No. 50-188-5453