Learn More
Aviva Systems Biology Corp MYEF2 Antibody - N-terminal region (ARP32065_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32065_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYEF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MYEF2. Alternate names: FLJ11213, HsT18564, KIAA1341, MEF-2, MGC87325, MST156, MSTP156ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54440