Learn More
Aviva Systems Biology Corp MYEF2 Antibody - middle region (ARP32738_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32738_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYEF2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human MYEF2. Alternate names: MEF-2, MST156, MSTP156, HsT18564ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-52870