Learn More
Enzo Life Sciences Myelin basic protein (human) (high purity) (1mg)
SDP
Supplier:  Enzo Life Sciences ALX200606M001
Encompass_Preferred
Alternative name: MBP. Concentration: 1mg/ml after reconstitution. Purity: ≥95% (SDS-PAGE; Immunoblot). MW: MBP contains 14.2kDa (13.0 minor) and 18.5kDa (17.2 minor) isoforms as well as their dimers (13.5% SDS-PAGE, non-reducing conditions). Appearance: White to off-white powder. Formulation: Lyophilized. No preservatives added. Source: Purified from human brain. Long Term Storage: -20°C. Use/Stability: Stable for 12 months at -20°C.
Catalog No. 50-200-7883