Learn More
Cell Signaling Technology Myelin Basic Protein Matched Antibody Pair, MBP, Unconjugated, Human, Mouse, Rat, 1 Kit
SDP
Catalog No. 503134075
Encompass
The Myelin Basic Protein Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms requiring antibody pairs with specialized labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for applications such as MSD, Quanterix Simoa, Alpha Technology, and Luminex. The antibodies are provided in a BSA and azide-free formulation, ensuring compatibility with custom conjugation strategies, including fluorophores, lanthanides, biotin, and beads.
  • Formulated in 1X PBS (pH 7.8) without BSA or azide
  • Validated for reactivity with human, mouse, and rat species
  • Ideal for diverse immunoassay platforms including ELISA, HTRF, and AlphaLISA
  • Capture and detection antibodies bind to non-overlapping epitopes
  • Store at -20°C
Catalog No. 50-313-4075 Supplier Cell Signaling Technology Supplier No. 74948S
The Myelin Basic Protein Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms requiring antibody pairs with specialized labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for applications such as MSD, Quanterix Simoa, Alpha Technology, and Luminex. The antibodies are provided in a BSA and azide-free formulation, ensuring compatibility with custom conjugation strategies, including fluorophores, lanthanides, biotin, and beads.
  • Formulated in 1X PBS (pH 7.8) without BSA or azide
  • Validated for reactivity with human, mouse, and rat species
  • Ideal for diverse immunoassay platforms including ELISA, HTRF, and AlphaLISA
  • Capture and detection antibodies bind to non-overlapping epitopes
  • Store at -20°C