Learn More
Medchemexpress LLC HY-P1048 1mg Medchemexpress, Myelin Basic Protein (MBP) (68-82), guinea pig CAS:98474-59-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP10481MG
Encompass_Preferred
Medchemexpress, HY-P1048 1mg Myelin Basic Protein (MBP) (68-82), guinea pig CAS:98474-59-0 Myelin Basic Protein (MBP) (68-82), guinea pig is a fragment of myelin basic protein (MBP). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4149