Learn More
Medchemexpress LLC HY-P1240A 5mg , Myelin Oligodendrocyte Glycoprotein Peptide (35-55), mouse, rat (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1240A5MG
Encompass_Preferred
HY-P1240A 5mg Myelin Oligodendrocyte Glycoprotein Peptide (35-55), mouse, rat (TFA) CAS: Myelin Oligodendrocyte Glycoprotein Peptide (35-55), mouse, rat (MOG (35-55)) TFA is a minor component of CNS myelin. Myelin Oligodendrocyte Glycoprotein Peptide (35-55), mouse, rat TFA has encephalitogenic activity and induces T cell proliferative. Myelin Oligodendrocyte Glycoprotein Peptide (35-55), mouse, rat TFA induces Th1 cytokine response as well as relatively high levels of IgG antibodies. Myelin Oligodendrocyte Glycoprotein Peptide (35-55), mouse, rat TFA produces a relapsing-remitting neurological disease with extensive plaque-like demyelination. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1207