Learn More
Bachem Myelin Oligodendrocyte Glycoprotein (35-55) (human) Trifluoroacetate
SDP
Supplier:  Bachem 4042037.1000
Encompass_Preferred
Myelin Oligodendrocyte Glycoprotein (35-55) (human) Trifluoroacetate, H-Met-Glu-Val-Gly-Trp-Tyr-Arg-Pro-Pro-Phe-Ser-Arg-Val-Val-His-Leu-Tyr-Arg-Asn-Gly-Lys-OH, 163158-19-8, C120H179N35O28S, Mr 2592.03, 1mg. MOG (35-55) (human). In contrast to the encephalitogenic rat homolog, myelin oligodendrocyte glycoprotein (35-55) (human) does not appear to be a major T-cell epitope in multiple sclerosis propably due to its lower amount of helical structure formed when dissolved in micellar sodium dodecyl sulfate, a membrane mimicking solvent.
Catalog No. 50-260-006