Learn More
Medchemexpress LLC HY-P1240A 1mg Medchemexpress, Myelin Oligodendrocyte Glycoprotein Peptide (35-55), mouse, rat (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1240A1MG
Encompass_Preferred
Medchemexpress, HY-P1240A 1mg Myelin Oligodendrocyte Glycoprotein Peptide (35-55), mouse, rat (TFA) CAS: Myelin Oligodendrocyte Glycoprotein Peptide (35-55), mouse, rat (TFA) is a minor component of CNS myelin. Myelin Oligodendrocyte Glycoprotein Peptide (35-55), mouse, rat (TFA) produces a relapsing-remitting neurological disease with extensive plaque-like demyelination. Myelin Oligodendrocyte Glycoprotein Peptide (35-55), mouse, rat (TFA) induces strong T and B cell responses and is highly encephalitogenic[3]98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0528