Learn More
CPC Scientific H-Met-Glu-Val-Gly-Trp-Tyr-Arg-Ser-Pro-Phe-Ser-Arg-Val-Val-His-Leu-Tyr-Arg-Asn-Gly-Lys-OH 5MG
SDP
Supplier:  CPC Scientific MOG001B
Encompass
Myelin Oligodendrocyte Glycoprotein peptidesSEQUENCE: H-Met-Glu-Val-Gly-Trp-Tyr-Arg-Ser-Pro-Phe-Ser-Arg-Val-Val-His-Leu-Tyr-Arg-Asn-Gly-Lys-OHONE-LETTER SEQUENCE: MEVGWYRSPFSRVVHLYRNGKMOLECULAR FORMULA: C118H177N35O29S1MOLECULAR WEIGHT: 2581.99STORAGE CONDITIONS:-20 5 CCAS REGISTRY NUMBER: [149635-73-4]SYNONYMS: MOG 35-55 (mouse, rat)RESEARCH AREA: Multiple SclerosisREFERENCES:PRODUCT CITATIONS:Le Moan, Natacha, et al. Eneuro 2.2 (2015): ENEURO-0050. Dupree, Jeffrey L., et al. Journal of Neurochemistry 134.2 (2015): 302-314. Orozco, Juan L. Hancke, and Rafael Burgos. U.S. Patent No. 9,060,994. 23 Jun. 2015.
Catalog No. 50-210-9814