Learn More
CPC Scientific H-His-Cys-Leu-Gly-Lys-Trp-Leu-Gly-His-Pro-Asp-Lys-Phe-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific MBP004B
Encompass
This peptide can induce autoimmune encephalomyelitis in mice for studying multiple sclerosis.ONE-LETTER SEQUENCE: HCLGKWLGHPDKFMOLECULAR FORMULA: C72H104N20O16S1MOLECULAR WEIGHT:1537.81STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [131334-43-5], SYNONYMS: PLP (139-151)RESEARCH AREA: Multiple SclerosisREFERENCES: S.Youssef et al., Nature, 420, 78 (2002); Gonzalez-Rey, E. et al. Am. J. Pathol. 168, 1179 (2006)
Catalog No. 50-210-9712