Learn More
Cayman Chemical Myeln BasIc protin PeptID 10mg
SDP
Catalog No. 502915444
Encompass_Preferred
An MBP antagonist; inhibits the binding of human MBP (85-99) to human HLA-DR2 in a concentration-dependent manner; inhibits the production of IL-2 in T cell hybridoma cells expressing an MBP (85-99)-specific T cell receptor derived from a patient with multiple sclerosis; suppresses MBP (85-99)-induced EAE onset and reduces EAE symptom severity in double transgenic humanized mice expressing human HLA-DR2 and MBP (85-99)-specific T cell receptor antigen in a mouse model of multiple sclerosis at 100 UG/animal
Catalog No. 50-291-5444 Supplier Cayman Chemical Supplier No. 3696410 MG
An MBP antagonist; inhibits the binding of human MBP (85-99) to human HLA-DR2 in a concentration-dependent manner; inhibits the production of IL-2 in T cell hybridoma cells expressing an MBP (85-99)-specific T cell receptor derived from a patient with multiple sclerosis; suppresses MBP (85-99)-induced EAE onset and reduces EAE symptom severity in double transgenic humanized mice expressing human HLA-DR2 and MBP (85-99)-specific T cell receptor antigen in a mouse model of multiple sclerosis at 100 UG/animal