Learn More
Cayman Chemical Myelin Basic Protein Peptide (mouse, bovine) (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502909676
Encompass_Preferred
A peptide substrate for PKC; selective for PKC over PKA, CK1, CK2, phosphorylase kinase, and CaMKII; has been used to monitor PKC activity in primary human renal artery smooth muscle cells
Catalog No. 50-290-9676 Supplier Cayman Chemical Supplier No. 3676110 MG
A peptide substrate for PKC; selective for PKC over PKA, CK1, CK2, phosphorylase kinase, and CaMKII; has been used to monitor PKC activity in primary human renal artery smooth muscle cells