Learn More
STEMCELL Technologies StemSpan™ Myeloid Expansion Supplement II (100X), Size: 1 mL
SDP
Catalog No. NC1469669
Encompass
StemSpan™ Myeloid Expansion Supplement II (100X) contains a combination of recombinant human cytokines formulated to selectively promote the expansion and differentiation of human myeloid progenitor cells, generating large numbers of CD14+ monocytes from CD34+ cells isolated from human cord blood (CB) samples.StemSpan™ Myeloid Expansion Supplement II (100X) is intended for use in combination with any of the following StemSpan™ media:• StemSpan™ SFEM (Catalog #09600)• StemSpan™ SFEM II (Catalog #09605)• StemSpan™-XF (Catalog #100-0073)• StemSpan™-AOF (Catalog #100-0130)When added to serum-free medium, StemSpan™ Myeloid Expansion Supplement II typically promotes the production of hundreds of CD14+ monocytes per input CD34+ cell in 14-day liquid cultures initiated with CD34+ human CB cells. See data tab for more details.
This item is not returnable. View return policy
Catalog No. NC1469669 Supplier STEMCELL Technologies Supplier No. 02694
StemSpan™ Myeloid Expansion Supplement II (100X) contains a combination of recombinant human cytokines formulated to selectively promote the expansion and differentiation of human myeloid progenitor cells, generating large numbers of CD14+ monocytes from CD34+ cells isolated from human cord blood (CB) samples.StemSpan™ Myeloid Expansion Supplement II (100X) is intended for use in combination with any of the following StemSpan™ media:• StemSpan™ SFEM (Catalog #09600)• StemSpan™ SFEM II (Catalog #09605)• StemSpan™-XF (Catalog #100-0073)• StemSpan™-AOF (Catalog #100-0130)When added to serum-free medium, StemSpan™ Myeloid Expansion Supplement II typically promotes the production of hundreds of CD14+ monocytes per input CD34+ cell in 14-day liquid cultures initiated with CD34+ human CB cells. See data tab for more details.