Learn More
CPC Scientific H-Leu-Val-Val-Tyr-Pro-Trp-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific OSTP005A
Encompass
MP-2 capable of restoring the T-cell phenotype altered by HL-60 CM. The MP-2 recovery effect is connected with its influence on the CD3 and CD4 antigen expression on T lymphocytes.SEQUENCE: H-Leu-Val-Val-Tyr-Pro-Trp-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: LVVYPWMOLECULAR FORMULA: C41H57N7O8MOLECULAR WEIGHT:775.95STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: OsteoporosisREFERENCES:L.A. Strelkov et al., Immunology Letters, 50, 143 (1996)SKU(s): OSTP-005A, OSTP-005B
Catalog No. 50-211-0007