Learn More
Diagnostic Biosystems MYELOPEROXIDASE Myeloperoxidase
Catalog No. 502407396
Encompass_Preferred
This antibody reacts with human myeloperoxidase. It stains granules of neutrophils, granulocytesin spleen, bone marrow, tonsil and blood smears.
Catalog No. 50-240-7396 Supplier Diagnostic Biosystems Supplier No. PDR049
This antibody reacts with human myeloperoxidase. It stains granules of neutrophils, granulocytesin spleen, bone marrow, tonsil and blood smears.