Learn More
Cell Signaling Technology Myeloperoxidase (E2Z8J) Rabbit Monoclonal Antibody, Human and Mouse Reactivity, 100 μL
SDP
Catalog No. NC2739655
Encompass
Myeloperoxidase (E2Z8J) Rabbit Monoclonal Antibody recognizes endogenous levels of total myeloperoxidase protein. This antibody detects the full-length and heavy chain subunits of human and mouse myeloperoxidase. It is generated by immunizing animals with a synthetic peptide corresponding to residues near the carboxy terminus of human myeloperoxidase protein.
  • Recognizes human and mouse myeloperoxidase
  • Validated for western blotting, immunofluorescence, and flow cytometry applications
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide
  • Store at −20°C; do not aliquot
Catalog No. NC2739655 Supplier Cell Signaling Technology Supplier No. 15178S
Myeloperoxidase (E2Z8J) Rabbit Monoclonal Antibody recognizes endogenous levels of total myeloperoxidase protein. This antibody detects the full-length and heavy chain subunits of human and mouse myeloperoxidase. It is generated by immunizing animals with a synthetic peptide corresponding to residues near the carboxy terminus of human myeloperoxidase protein.
  • Recognizes human and mouse myeloperoxidase
  • Validated for western blotting, immunofluorescence, and flow cytometry applications
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide
  • Store at −20°C; do not aliquot