Learn More
Abcam Myelopero x idase Inhibitor, MPO inhibitor, 100MG
Catalog No. 502706580
Encompass_Preferred
MW 151.17 Da, Purity >95%. Potent, irreversible and specific MPO inhibitor (IC₅₀ = 300 nM). Benzoic acid analog. No inhibition of catalase or glutathione peroxidase activity. Inhibits apoptosis in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6580 Supplier Abcam Supplier No. AB141482100MG
MW 151.17 Da, Purity >95%. Potent, irreversible and specific MPO inhibitor (IC₅₀ = 300 nM). Benzoic acid analog. No inhibition of catalase or glutathione peroxidase activity. Inhibits apoptosis in vitro.
The product is subject to the following: Abcam Restricted Use Statement