Learn More
Cayman Chemical Myeloperoxidase Monoclonal Antibody (Clone 5F6), 1 ea
SDP
Catalog No. 502972599
Encompass_Preferred
Host: Mouse • Species Reactivity: (+) Human, mouse • Application: IF
Catalog No. 50-297-2599 Supplier Cayman Chemical Supplier No. 156401 EA
Host: Mouse • Species Reactivity: (+) Human, mouse • Application: IF