Learn More
Athens Research And Technology Inc MYELOPEROXIDASE NEUTROPHIL 100
Catalog No. 50489881
Encompass_Preferred
Myeloperoxidase, Human Neutrophil: 100 UG; Lyophilized from 50 mM Na acetate, pH 6.0 w/ 100 mM NaCl; MW 150,000; EC= 1.45; Activity: 180-220 units per mg protein; Purity: > =95% by SDS-PAGE. Native; Purified from whole blood.
Catalog No. 50-489-881 Supplier Athens Research And Technology Inc Supplier No. 1614130000100UG
Myeloperoxidase, Human Neutrophil: 100 UG; Lyophilized from 50 mM Na acetate, pH 6.0 w/ 100 mM NaCl; MW 150,000; EC= 1.45; Activity: 180-220 units per mg protein; Purity: > =95% by SDS-PAGE. Native; Purified from whole blood.