Learn More
Athens Research And Technology Inc MYELOPEROXIDASE NEUTROPHIL 1MG
Catalog No. 50489883
Encompass_Preferred
Myeloperoxidase, Human Neutrophil: 1 MG; Lyophilized from 50 mM Na acetate, pH 6.0 with 100 mM NaCl; MW 150,000; EC= 1.45; Activity: 180-220 units/mg protein; Purity: > =95% by SDS-PAGE. Native; Purified from whole blood in the Athens Research & Technology laboratory. Molecular Biology Reagent
Catalog No. 50-489-883 Supplier Athens Research And Technology Inc Supplier No. 16141300001MG
Myeloperoxidase, Human Neutrophil: 1 MG; Lyophilized from 50 mM Na acetate, pH 6.0 with 100 mM NaCl; MW 150,000; EC= 1.45; Activity: 180-220 units/mg protein; Purity: > =95% by SDS-PAGE. Native; Purified from whole blood in the Athens Research & Technology laboratory. Molecular Biology Reagent