Learn More
Roche Diagnostics Myeloperoxidase Rabbit Polyclonal Antibody, 50 tests
Catalog No. NC2895018 Shop All Roche Diagnostics Products
Encompass
Anti-Myeloperoxidase Polyclonal Primary Antibody is intended for laboratory use in the detection of the myeloperoxidase protein in formalin-fixed, paraffin-embedded human tissue stained in qualitative immunohistochemistry (IHC) on BenchMark IHC/ISH instruments. This product should be interpreted by a qualified pathologist in conjunction with histological examination, relevant clinical information, and proper controls. This antibody is intended for in vitro diagnostic (IVD) use.
Catalog No. NC2895018 Supplier Roche Diagnostics Supplier No. 5267692001
Anti-Myeloperoxidase Polyclonal Primary Antibody is intended for laboratory use in the detection of the myeloperoxidase protein in formalin-fixed, paraffin-embedded human tissue stained in qualitative immunohistochemistry (IHC) on BenchMark IHC/ISH instruments. This product should be interpreted by a qualified pathologist in conjunction with histological examination, relevant clinical information, and proper controls. This antibody is intended for in vitro diagnostic (IVD) use.