Learn More
Medchemexpress LLC MYF-01-37 | 2416417-65-5 | 99.4% | 298.30 | 100 MG
SDP
Catalog No. 5000349499
Encompass_Preferred
MYF-01-37 is a covalent TEAD inhibitor that targets Cys380. It exhibits reversible inhibition on YAP/TEAD interaction and is intended for research use only, not for sale to patients. In vitro studies show that MYF-01-37 inhibits direct YAP/TEAD interaction in HEK 293T cells and reduces canonical YAP target gene CTGF expression in PC-9 cells.
  • Covalent TEAD inhibitor targeting Cys380
  • Reversible inhibition on YAP/TEAD interaction
  • Inhibits direct YAP/TEAD interaction in HEK 293T cells
  • Reduces canonical YAP target gene CTGF expression in PC-9 cells
  • Minimal impact on cell viability of several EGFR-mutant NCSLC cell lines
  • Decreases dormant cells when combined with other treatments
Catalog No. 50-003-49499 Supplier Medchemexpress LLC Supplier No. HY139603100MG
MYF-01-37 is a covalent TEAD inhibitor that targets Cys380. It exhibits reversible inhibition on YAP/TEAD interaction and is intended for research use only, not for sale to patients. In vitro studies show that MYF-01-37 inhibits direct YAP/TEAD interaction in HEK 293T cells and reduces canonical YAP target gene CTGF expression in PC-9 cells.
  • Covalent TEAD inhibitor targeting Cys380
  • Reversible inhibition on YAP/TEAD interaction
  • Inhibits direct YAP/TEAD interaction in HEK 293T cells
  • Reduces canonical YAP target gene CTGF expression in PC-9 cells
  • Minimal impact on cell viability of several EGFR-mutant NCSLC cell lines
  • Decreases dormant cells when combined with other treatments