Learn More
Medchemexpress LLC Myf-01-37 | 2416417-65-5 | 99.44% | 298.30 | 50 MG
SDP
Catalog No. 5000350109
Encompass_Preferred
MYF-01-37 is a covalent TEAD inhibitor that targets Cys380 and demonstrates a reversible inhibition of the YAP/TEAD interaction. It has minimal impact on the cell viability of several EGFR-mutant NCSLC cell lines and, when combined with osimertinib and trametinib, leads to a significant decrease in dormant cells. This product is intended for research use only.
  • Inhibits direct YAP/TEAD interaction in HEK 293T cells
  • Reduces canonical YAP target gene CTGF expression in PC-9 cells
  • Minimal impact on cell viability of EGFR-mutant NCSLC cell lines
  • Decreases dormant cells when combined with OT
Catalog No. 50-003-50109 Supplier Medchemexpress LLC Supplier No. HY13960350MG
Add to Cart
Add to Cart
MYF-01-37 is a covalent TEAD inhibitor that targets Cys380 and demonstrates a reversible inhibition of the YAP/TEAD interaction. It has minimal impact on the cell viability of several EGFR-mutant NCSLC cell lines and, when combined with osimertinib and trametinib, leads to a significant decrease in dormant cells. This product is intended for research use only.
  • Inhibits direct YAP/TEAD interaction in HEK 293T cells
  • Reduces canonical YAP target gene CTGF expression in PC-9 cells
  • Minimal impact on cell viability of EGFR-mutant NCSLC cell lines
  • Decreases dormant cells when combined with OT