Learn More
TARGETMOL CHEMICALS INC MYF-01-37 25MG
SDP
Catalog No. 502580957
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MYF-01-37 (1-[3-Methyl-3-[3-(trifluoromethyl)anilino]pyrrolidin-1-yl]prop-2-en-1-one) is a novel covalent TEAD inhibitor. purity: 99%
Catalog No. 50-258-0957 Supplier TARGETMOL CHEMICALS INC Supplier No. T2237225MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MYF-01-37 (1-[3-Methyl-3-[3-(trifluoromethyl)anilino]pyrrolidin-1-yl]prop-2-en-1-one) is a novel covalent TEAD inhibitor. purity: 99%