Learn More
Selleck Chemical LLC MYF-01-37 S8950-25MG
SDP
Catalog No. 5000024223
Encompass_Preferred
MYF-01-37 is a novel covalent inhibitor of TEAD targeting Cys380 when incubated with the TEAD2 protein
Catalog No. 50-000-24223 Supplier Selleck Chemical LLC Supplier No. S895025MG
MYF-01-37 is a novel covalent inhibitor of TEAD targeting Cys380 when incubated with the TEAD2 protein