Learn More
Selleck Chemical LLC MYF-01-37 5mg
SDP
Supplier:  Selleck Chemical LLC S89505MG
Encompass_Preferred
MYF-01-37 is a novel covalent inhibitor of TEAD targeting Cys380 when incubated with the TEAD2 protein. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3451