Learn More
Medchemexpress LLC MYF-03-176 | 2857937-59-6 | 99.72% | 410.37 | 25 MG
SDP
Catalog No. 5000363699
Encompass_Preferred
MYF-03-176 is an orally active inhibitor of TEAD, suppressing TEAD transcriptional activity with an IC50 of 11 nM. It inhibits hippo signaling defective malignant pleural mesothelioma (MPM) cells and demonstrates strong antitumor efficacy in an MPM mouse xenograft model. The IC50 values for TEAD1, TEAD3, and TEAD4 are 47, 32, and 71 nM, respectively.
  • Orally active inhibitor of TEAD
  • Suppresses TEAD transcriptional activity with an IC50 of 11 nM
  • Inhibits hippo signaling defective malignant pleural mesothelioma (MPM) cells
  • Demonstrates strong antitumor efficacy in MPM mouse xenograft model
Catalog No. 50-003-63699 Supplier Medchemexpress LLC Supplier No. 25MGHY147136
MYF-03-176 is an orally active inhibitor of TEAD, suppressing TEAD transcriptional activity with an IC50 of 11 nM. It inhibits hippo signaling defective malignant pleural mesothelioma (MPM) cells and demonstrates strong antitumor efficacy in an MPM mouse xenograft model. The IC50 values for TEAD1, TEAD3, and TEAD4 are 47, 32, and 71 nM, respectively.
  • Orally active inhibitor of TEAD
  • Suppresses TEAD transcriptional activity with an IC50 of 11 nM
  • Inhibits hippo signaling defective malignant pleural mesothelioma (MPM) cells
  • Demonstrates strong antitumor efficacy in MPM mouse xenograft model