Learn More
Aviva Systems Biology Corp MYF6 Antibody - N-terminal region (ARP32135_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32135_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYF6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MYF6. Alternate names: MRF4, CNM3, myf-6, bHLHc4ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-54459