Learn More
Aviva Systems Biology Corp MYF6 Antibody - middle region (ARP31409_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31409_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYF6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MYF6. Alternate names: MRF4, CNM3, myf-6, bHLHc4ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-54211