Learn More
ABclonal Technology MYF6 Rabbit pAb
SDP
Catalog No. 5000105308
Encompass_Preferred
The protein encoded by this gene is a probable basic helix-loop-helix (bHLH) DNA binding protein involved in muscle differentiation. The encoded protein likely acts as a heterodimer with another bHLH protein. Defects in this gene are a cause of autosomal dominant centronuclear myopathy (ADCNM).
Catalog No. 50-001-05308 Supplier ABclonal Technology Supplier No. A15291500UL
The protein encoded by this gene is a probable basic helix-loop-helix (bHLH) DNA binding protein involved in muscle differentiation. The encoded protein likely acts as a heterodimer with another bHLH protein. Defects in this gene are a cause of autosomal dominant centronuclear myopathy (ADCNM).