Learn More
Meridian Life Science MyFi DNA Polymerase, 250 units
SDP
Catalog No. NC1595388
Encompass
Unique blend of highly-efficient MyTaq HS DNA Polymerase and a proprietary proofreading enzyme that combine to give increased target affinity for use with challenging templates and inhibitor-rich samples.
Catalog No. NC1595388 Supplier Meridian Life Science Supplier No. BIO21117
Unique blend of highly-efficient MyTaq HS DNA Polymerase and a proprietary proofreading enzyme that combine to give increased target affinity for use with challenging templates and inhibitor-rich samples.