Learn More
Research Products International Corp myFuge 12 Micro-Centrifuge, for 12 Microtubes and/or 4 PCR Strips
SDP
Catalog No. 50550338
Encompass
Unlike traditional mini centrifuges, the new MyFuge 12 eliminates the need to change rotors when switching between microtubes and PCR strips. The unique COMBI-Rotor, which is included, is all that is required for running 12 microtubes and/or 4 PCR strips. With a fixed speed that produces 2,000 xg, this centrifuge is perfect for quick spin downs. Simply close the lid and the unit quickly ramps up to 5500 rpm. Open the lid, and the rotor quickly decelerates for removal of samples. At just 5.5 inches wide and less than 4.5 inches tall, the MyFuge 12 truly is a personal centrifuge with unmatched capacity and flexibility.
Catalog No. 50-550-338 Supplier Research Products International Corp Supplier No. 400755
Unlike traditional mini centrifuges, the new MyFuge 12 eliminates the need to change rotors when switching between microtubes and PCR strips. The unique COMBI-Rotor, which is included, is all that is required for running 12 microtubes and/or 4 PCR strips. With a fixed speed that produces 2,000 xg, this centrifuge is perfect for quick spin downs. Simply close the lid and the unit quickly ramps up to 5500 rpm. Open the lid, and the rotor quickly decelerates for removal of samples. At just 5.5 inches wide and less than 4.5 inches tall, the MyFuge 12 truly is a personal centrifuge with unmatched capacity and flexibility.