Learn More
Andwin Scientific MYFUGE 12 MINI CENTRIFUGE EA
SDP
Catalog No. 50999380
Encompass_Preferred
The MyFuge 12 Mini Centrifuge removes the need of changing rotors when switching between microtubes and PCR strips. This product is sold with the COMBI-Rotor, which is all that is needed for running 12 microtubes and 4 PCR strips at the same time. The Mini Centrifuge has twice the capacity of traditional mini centrifuges and it begins and stops the operation with the opening and closing of the lid.
Catalog No. 50-999-380 Supplier Andwin Scientific Supplier No. C1012
The MyFuge 12 Mini Centrifuge removes the need of changing rotors when switching between microtubes and PCR strips. This product is sold with the COMBI-Rotor, which is all that is needed for running 12 microtubes and 4 PCR strips at the same time. The Mini Centrifuge has twice the capacity of traditional mini centrifuges and it begins and stops the operation with the opening and closing of the lid.