Learn More
Alkali Scientific Alkali Scientific™ MyFuge™ 12 Plus C2212 Mini Centrifuge w/COMBI-Rotor
SDP
Catalog No. 503029596
Encompass_Preferred
COMBI-Rotor for tubes and strips Digital control up to 7,000 rpm / 2,739 xg! Twice the capacity of traditional mini centrifuges Near silent operation Timer up to 99 mins. Alkali Scientific™'s (C2212) unique rotor design of the new MyFuge 12 Plus eliminates the need to switch rotors (or use a separate centrifuge) when changing between microtubes and PCR strips. The included, COMBI-Rotor allows for simultaneous centrifugation of up to 12 microtubes and/or 4 PCR strips. In addition, the MyFuge 12 Plus features a large digital display with adjustable speed (up to 7,000 rpm) and a digital timer. With built in rcf conversion, speed can be set/displayed in rpm or rcf. The MyFuge 12 Plus combines power with intuitive functionality making it the perfect companion in any laboratory.
Catalog No. 50-302-9596 Supplier Alkali Scientific Supplier No. RS2212
COMBI-Rotor for tubes and strips Digital control up to 7,000 rpm / 2,739 xg! Twice the capacity of traditional mini centrifuges Near silent operation Timer up to 99 mins. Alkali Scientific™'s (C2212) unique rotor design of the new MyFuge 12 Plus eliminates the need to switch rotors (or use a separate centrifuge) when changing between microtubes and PCR strips. The included, COMBI-Rotor allows for simultaneous centrifugation of up to 12 microtubes and/or 4 PCR strips. In addition, the MyFuge 12 Plus features a large digital display with adjustable speed (up to 7,000 rpm) and a digital timer. With built in rcf conversion, speed can be set/displayed in rpm or rcf. The MyFuge 12 Plus combines power with intuitive functionality making it the perfect companion in any laboratory.