Learn More
Andwin Scientific MYFUGE BLUELID MICROCENTRIFUGE
SDP
Catalog No. NC0211385
Encompass_Preferred
The Mini Centrifuge is made to work for the centrifugation of microcentrifuge tubes and PCR strips. This product is designed with a quick-release rotor adapter, which does not require extra tools, that can hold either an 8-Place tube or 4x8 PCR strip
Catalog No. NC0211385 Supplier Andwin Scientific Supplier No. C1008B
Add to Cart
Add to Cart
The Mini Centrifuge is made to work for the centrifugation of microcentrifuge tubes and PCR strips. This product is designed with a quick-release rotor adapter, which does not require extra tools, that can hold either an 8-Place tube or 4x8 PCR strip