Learn More
Braintree Scientific MyFuge Mini Centrifuge- Red
SDP
Catalog No. 501954467
Encompass
MyFuge Mini Centrifuge. Eight position microtube rotor. Storage compartment for PCR rotor. Near silent operation. Starts and stops in just seconds. Conserves valuable bench space. Specifications: Speed: 6,000 rpm/2,000 xg; Dimensions: 4.5" x 5.9" x 4.5", 11.4cm x 15cm x 11cm; Capacity: 8 x 1.5/2.0mI tubes, 16 x .2ml PCR tubes, 2 x .2ml PCR strips. MyFUGE™ mini centrifuge is a personal centrifuge that fits in the palm of your hand. Ideal for quick spin downs of microtubes and PCR tubes, MyFuge is extremely easy to use. Simply close lid and rotor instantly reaches 6,000 rpm. Open lid, and rotor quickly decelerates to a stop for removing samples. An increased capacity eight position microtube rotor is included and comes installed. A second rotor for .2ml PCR tubes and strips is also included and stored with rotor removal key in a convenient compartment on bottom of the centrifuge. All include microtube rotor, PCR rotor, .5ml and .2ml adapter.
Catalog No. 50-195-4467 Supplier Braintree Scientific Supplier No. C1008R
MyFuge Mini Centrifuge. Eight position microtube rotor. Storage compartment for PCR rotor. Near silent operation. Starts and stops in just seconds. Conserves valuable bench space. Specifications: Speed: 6,000 rpm/2,000 xg; Dimensions: 4.5" x 5.9" x 4.5", 11.4cm x 15cm x 11cm; Capacity: 8 x 1.5/2.0mI tubes, 16 x .2ml PCR tubes, 2 x .2ml PCR strips. MyFUGE™ mini centrifuge is a personal centrifuge that fits in the palm of your hand. Ideal for quick spin downs of microtubes and PCR tubes, MyFuge is extremely easy to use. Simply close lid and rotor instantly reaches 6,000 rpm. Open lid, and rotor quickly decelerates to a stop for removing samples. An increased capacity eight position microtube rotor is included and comes installed. A second rotor for .2ml PCR tubes and strips is also included and stored with rotor removal key in a convenient compartment on bottom of the centrifuge. All include microtube rotor, PCR rotor, .5ml and .2ml adapter.