Learn More
Braintree Scientific MyFuge12 Mini Centrifugew/COMBI-Rotor for microtubes & PCR
SDP
Catalog No. 501954468
Encompass
MyFuge12 Mini Centrifuge. COMBI-Rotor™ for tubes and strips. Twice the capacity of traditional mini centrifuges. Near silent operation. Starts and stops with opening/closing of lid. Unlike traditional mini centrifuges, the MyFuge 12 eliminates the need to change rotors when switching between microtubes and PCR strips. The included, unique COMBI-Rotor is all that is required for running 12 microtubes and 4 PCR strips simultaneously. With a fixed speed that produces 2,000 xg, this centrifuge is perfect for quick spin downs. Simply close lid and unit quickly ramps up to 5500 rpm. Open lid, and rotor quickly decelerates for removal of samples. At just 5.5" wide and less than 4.5" tall, MyFuge 12 truly is a personal centrifuge with unmatched capacity and flexibiilty. Specifications: Speed: 5,500 rpm/2,000 xg; Capacity: 12 x 1.5/2.0ml tubes, 32 x .2ml PCR tubes; 4 x PCR strips (8 x .2ml); Dimensions: 5.5" x 7.9" x 4.4"/14 cm x 20cm x 11.2cm.
Catalog No. 50-195-4468 Supplier Braintree Scientific Supplier No. C1012
MyFuge12 Mini Centrifuge. COMBI-Rotor™ for tubes and strips. Twice the capacity of traditional mini centrifuges. Near silent operation. Starts and stops with opening/closing of lid. Unlike traditional mini centrifuges, the MyFuge 12 eliminates the need to change rotors when switching between microtubes and PCR strips. The included, unique COMBI-Rotor is all that is required for running 12 microtubes and 4 PCR strips simultaneously. With a fixed speed that produces 2,000 xg, this centrifuge is perfect for quick spin downs. Simply close lid and unit quickly ramps up to 5500 rpm. Open lid, and rotor quickly decelerates for removal of samples. At just 5.5" wide and less than 4.5" tall, MyFuge 12 truly is a personal centrifuge with unmatched capacity and flexibiilty. Specifications: Speed: 5,500 rpm/2,000 xg; Capacity: 12 x 1.5/2.0ml tubes, 32 x .2ml PCR tubes; 4 x PCR strips (8 x .2ml); Dimensions: 5.5" x 7.9" x 4.4"/14 cm x 20cm x 11.2cm.