Learn More
Sigma Aldrich Fine Chemicals Biosciences MyfugeTM 12 mini centrifug1EA
Catalog No. 502905655
Encompass
Myfuge 12 mini centrifuge eliminates the need to change rotors when switching between microtubes and PCR strips. The included COMBI-Rotor is all that is required for running 12 microtubes and 4 PCR strips simultaneously.With a fixed speed that produces 2 000 x g this centrifuge is perfect for quick spin downs. Simply close the lid and the unit quickly ramps up to 5500 rpm. Open the lid and the rotor quickly decelerates for removal of samples.Capacity12 x 1.5/2.0 mL tubes32 x 0.2 mL PCR tubes4 x PCR strips (8 x 0.2 mL)
Catalog No. 50-290-5655 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6817171EA
Myfuge 12 mini centrifuge eliminates the need to change rotors when switching between microtubes and PCR strips. The included COMBI-Rotor is all that is required for running 12 microtubes and 4 PCR strips simultaneously.With a fixed speed that produces 2 000 x g this centrifuge is perfect for quick spin downs. Simply close the lid and the unit quickly ramps up to 5500 rpm. Open the lid and the rotor quickly decelerates for removal of samples.Capacity12 x 1.5/2.0 mL tubes32 x 0.2 mL PCR tubes4 x PCR strips (8 x 0.2 mL)