Learn More
Sigma Aldrich Fine Chemicals Biosciences MyfugeTM mini centrifuge b1EA
Catalog No. 502905692
Encompass
The Myfuge(TM) Mini centrifuge is a personal centrifuge that virtually fits in the palm of your hand. Ideal for quick spin downs of microtubes and PCR tubes the Myfuge is extremely easy to use. Simply close the lid and the rotor instantly reaches 6 000 rpm.Open the lid and the rotor quickly decelerates to a stop for removing samples. An increased capacity eight-position microtube rotor is included and comes installed.Also included is a second rotor for 0.2 mL PCR tubes and strips which is stored (together with the rotor removal key) in a convenient compartment on the bottom of the centrifuge. When the rotor is not in use storage in this compartment prevents loss or misplacement of the rotor.Eight position microtube rotorStorage compartment for PCR rotorSupplied with microtube rotor PCR rotor 0.5 mL and 2.0 mL adapters
Catalog No. 50-290-5692 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7638531EA
The Myfuge(TM) Mini centrifuge is a personal centrifuge that virtually fits in the palm of your hand. Ideal for quick spin downs of microtubes and PCR tubes the Myfuge is extremely easy to use. Simply close the lid and the rotor instantly reaches 6 000 rpm.Open the lid and the rotor quickly decelerates to a stop for removing samples. An increased capacity eight-position microtube rotor is included and comes installed.Also included is a second rotor for 0.2 mL PCR tubes and strips which is stored (together with the rotor removal key) in a convenient compartment on the bottom of the centrifuge. When the rotor is not in use storage in this compartment prevents loss or misplacement of the rotor.Eight position microtube rotorStorage compartment for PCR rotorSupplied with microtube rotor PCR rotor 0.5 mL and 2.0 mL adapters