Learn More
Alkali Scientific myGel InstaView Electrophoresis System, includes gel tank with orange filter, direct connect power supply, E1200-CS1 casting set, Mini SmartBlue Transilluminator and imaging enclosure
SDP
Catalog No. 502336029
Encompass_Preferred
Complete system with power supply The myGel InstaView comes complete with everything needed to cast, run and view mini electrophoresis gels, including an integrated power supply. One large (10.5x10cm) or small (10.5x6cm) gel is accommodated by the gel tank. Gels are easily poured using the supplied trays, combs and leak-proof casting stand. The power supply connects directly into the gel tank - no more leads to contend with. The connection is guaranteed to be right, every time. Output voltage can be adjusted to 35V, 50V or 100V and run time can be set for up to 99 minutes. A magnetic interlock between the gel tank and the lid prevents current from running through the system when the lid is not properly in place. Once the myGel InstaView System is set up, simply slide the mini LED transilluminator into place under the tank and at any time turn it on to view the run in real time. An array of blue LEDs emits bright blue light at a wavelength of 465nm for optimal excitation of c
Catalog No. 50-233-6029 Supplier Alkali Scientific Supplier No. ES1201
Complete system with power supply The myGel InstaView comes complete with everything needed to cast, run and view mini electrophoresis gels, including an integrated power supply. One large (10.5x10cm) or small (10.5x6cm) gel is accommodated by the gel tank. Gels are easily poured using the supplied trays, combs and leak-proof casting stand. The power supply connects directly into the gel tank - no more leads to contend with. The connection is guaranteed to be right, every time. Output voltage can be adjusted to 35V, 50V or 100V and run time can be set for up to 99 minutes. A magnetic interlock between the gel tank and the lid prevents current from running through the system when the lid is not properly in place. Once the myGel InstaView System is set up, simply slide the mini LED transilluminator into place under the tank and at any time turn it on to view the run in real time. An array of blue LEDs emits bright blue light at a wavelength of 465nm for optimal excitation of c