Learn More
Alkali Scientific myGel Mini Electrophoresis System, complete with power supply, two 10.5 x 6 cm and four 5 x 6 cm gel trays, casting stands for each size of gel tray and two double sided combs for each tray size
SDP
Catalog No. 502336056
Encompass_Preferred
The myGel™ Horizontal Electrophoresis System includes all the equipment you need to get up and running - gel tank, power supply and 2 complete casting sets. Ideal for personal use, small laboratories, or the classroom, this gel box/power supply combination is compact and easy to use.The myGel Mini power supply attaches directly to the gel box for a secure connection every time. Selectable parameters include 3 preset voltages - 35, 50 and 100V - and a 99 minute timer. All are set using the membrane keypad.The safety lid is vented to help to dissipate heat generated during electrophoresis, and features a viewing area above the gel. Current will only run through the sysem when the lid is properly in place on top of the gel box.Trays and casting stands for both 10.5 x 6 cm and 5 x 6 cm gels are included with the myGel Mini system, as are double sided combs. The smaller tooth combs are compatible with multichannel pipettes. Casting is quick and easy - just place the tray in
Catalog No. 50-233-6056 Supplier Alkali Scientific Supplier No. ES1101
The myGel™ Horizontal Electrophoresis System includes all the equipment you need to get up and running - gel tank, power supply and 2 complete casting sets. Ideal for personal use, small laboratories, or the classroom, this gel box/power supply combination is compact and easy to use.The myGel Mini power supply attaches directly to the gel box for a secure connection every time. Selectable parameters include 3 preset voltages - 35, 50 and 100V - and a 99 minute timer. All are set using the membrane keypad.The safety lid is vented to help to dissipate heat generated during electrophoresis, and features a viewing area above the gel. Current will only run through the sysem when the lid is properly in place on top of the gel box.Trays and casting stands for both 10.5 x 6 cm and 5 x 6 cm gels are included with the myGel Mini system, as are double sided combs. The smaller tooth combs are compatible with multichannel pipettes. Casting is quick and easy - just place the tray in