Learn More
Aviva Systems Biology Corp MYH10 Antibody - N-terminal region (ARP42270_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42270_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYH10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MYH10. Alternate names: MGC134913, MGC134914, NMMHCB, NMMHC-IIBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58169