Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4629MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: ID8. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Uterus or normal breast. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human uterus extract. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 205kDa (MHC-1), 200kDa (MHC-2).Chromosome: 16p13.11. Gene Symbol: MYH11. Gene ID: 4629. Ensemble: ENSG00000276480. Uniprot: XP_016878739.
Catalog No. 50-188-4985