Learn More
Aviva Systems Biology Corp MYH9 Antibody - middle region (ARP48072_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48072_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYH9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MYH9. Alternate names: DFNA17, EPSTS, FTNS, MGC104539, MHA, NMHC-II-A, NMMHCA, BDPLT6, NMMHC-IIAApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59713