Learn More
ABclonal Technology MYL2 Rabbit pAb
SDP
Catalog No. 5000106250
Encompass_Preferred
This gene encodes a major sarcomeric protein in mammalian striated muscle. The encoded protein plays a role in embryonic heart muscle structure and function, while phosphorylation of the encoded protein is involved in cardiac myosin cycling kinetics, torsion and function in adults. Mutations in this gene are associated with hypertrophic cardiomyopathy 10 and infant-onset myopathy.
Catalog No. 50-001-06250 Supplier ABclonal Technology Supplier No. A14188500UL
This gene encodes a major sarcomeric protein in mammalian striated muscle. The encoded protein plays a role in embryonic heart muscle structure and function, while phosphorylation of the encoded protein is involved in cardiac myosin cycling kinetics, torsion and function in adults. Mutations in this gene are associated with hypertrophic cardiomyopathy 10 and infant-onset myopathy.