Learn More
Aviva Systems Biology Corp MYL4 Antibody - N-terminal region (ARP56153_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56153_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYL4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human MYL4. Alternate names: ALC1, AMLC, GT1, PRO1957ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-286-120