Learn More
Aviva Systems Biology Corp MYL6 Antibody - N-terminal region (ARP57711_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57711_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYL6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MYL6. Alternate names: ESMLC, LC17-GI, LC17-NM, LC17A, LC17B, MLC1SM, MLC3NM, MLC3SM, LC17, MLC-3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-62137