Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ MyLab™ Custom Kit - Building DNA With Beads
SDPSCI_ED

Catalog No. S06465 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
6 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06465 6 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06465 Supplier EDVOTEK™ Supplier No. 1103

missing translation for 'validMainframeMsg'

Are you planning a virtual curriculum or looking for experiments ideal for distance learning? With the MyLab™ Custom Kits, you can easily plan your curriculum. Each kit comes with instructions and materials for 2-3 runs of each experiment.

  • Understand DNA base pairing
  • Model the structure of DNA
  • Explore palindromes, restriction enzymes, DNA synthesis, and genetic engineering
  • Includes instructions, 75 colored beads

Specifications

Grade 6 to 15
Age 12 to 20
For Use With (Application) At Home Education
Includes Instructions, 75 Colored Beads
Quantity 1 Kit
Description Building DNA With Beads
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.