Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ MyLab™ Custom Kit - Crime Scene DNA Fingerprinting
SDPSCI_ED

Catalog No. S06470 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
9 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S06470 9 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S06470 Supplier EDVOTEK™ Supplier No. 1230

missing translation for 'validMainframeMsg'

Are you planning a virtual curriculum or looking for experiments ideal for distance learning? With the MyLab™ Custom Kits, you can easily plan your curriculum. Each kit comes with instructions and materials for 2-3 runs of each experiment.

Forensic scientists use DNA fingerprinting to analyze crime scene DNA evidence and identify missing people. In this simulation of DNA fingerprinting, students will utilize paper chromatography to analyze the crime scene and suspect samples to identify 'whodunnit'.

  • Explore DNA analysis and its forensic applications
  • Learn about restriction enzymes, electrophoresis and chromatography
  • Analyze crime scene and suspect samples to determine ‘whodunnit’
  • Includes instructions, simulated DNA samples, chromatography paper, buffer, pipettes

Specifications

Grade 9 to 15
Age 14 to 20
For Use With (Application) At Home Education
Includes Instructions, Simulated DNA Samples, Chromatography Paper, Buffer, Pipettes
Quantity 1 Kit
Description Crime Scene DNA Fingerprinting
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.